missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human vWF-A2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF2764
This item is not returnable.
View return policy
Description
vWF-A2 Polyclonal specifically detects vWF-A2 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| vWF-A2 | |
| Polyclonal | |
| Unconjugated | |
| NP_000543 | |
| VWF | |
| E. coli-derived recombinant human vWF-A2 Asp1498-Val1665 Accession # NP_000543 | |
| 100 μg | |
| Primary | |
| Detects human vWF-A2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunoprecipitation 25 ug/mL | |
| F8VWF, VWD, VWF von Willebrand factor, vWFA2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7450 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction