missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VSTM2A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10037-100
This item is not returnable.
View return policy
Description
VSTM2A Monoclonal specifically detects VSTM2A in Human samples. It is validated for ELISA.
Specifications
| VSTM2A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2 | |
| Human embryonic kidney cell HEK293-derived human VSTM2A, Ser25-Phe244, Accession # Q8TAG5-2 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 1024718 | |
| ELISA | |
| Q8TAG5-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 222008 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction