missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VSIG3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £408.00
Specifications
| Antigen | VSIG3/IGSF11 |
|---|---|
| Clone | 973422 |
| Dilution | Flow Cytometry 0.25 ug/106 cells, Blockade of Receptor-ligand Interaction 0.2-2 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Blocking Assay, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780737
![]() |
R&D Systems
MAB92293-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701077
![]() |
R&D Systems
MAB92293-100 |
100 μg |
£408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSIG3/IGSF11 Monoclonal antibody specifically detects VSIG3/IGSF11 in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOFSpecifications
| VSIG3/IGSF11 | |
| Flow Cytometry 0.25 ug/106 cells, Blockade of Receptor-ligand Interaction 0.2-2 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein | |
| Mouse myeloma cell line NS0-derived recombinant human VSIG3, Met1-Gly245, Accession # Q5DX21 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 973422 | |
| Flow Cytometry, Blocking Assay, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q5DX21 | |
| 152404 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title