missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VSIG2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB51771G-100UG
This item is not returnable.
View return policy
Description
VSIG2 Monoclonal specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry.
Specifications
| VSIG2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| 2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
| Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession No. Q961Q7-1 | |
| 100 μg | |
| 23584.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 541522 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q961Q7-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human VSIG2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction