missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VPS52 (aa 581-677) Control Fragment Recombinant Protein

Product Code. 30182181
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182181

Brand: Invitrogen™ RP99253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61662 (PA5-61662. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that is similar to the yeast suppressor of actin mutations 2 gene. The yeast protein forms a subunit of the tetrameric Golgi-associated retrograde protein complex that is involved in vesicle trafficking from both early and late endosomes, back to the trans-Golgi network. This gene is located on chromosome 6 in a head-to-head orientation with the gene encoding ribosomal protein S18.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N1B4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6293
Name Human VPS52 (aa 581-677) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Are1; D130068D18; D430041K17Rik; dJ1033B10.5; DKFZp547I194; fb94c11; LOW QUALITY PROTEIN: vacuolar protein sorting-associated protein 52 homolog; RP5-1033B10; SAC2; SAC2 suppressor of actin mutations 2-like protein; SACM2L; tclw5; vacuolar protein sorting 52 (yeast); vacuolar protein sorting 52 homolog; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; vacuolar protein sorting-associated protein 52-like protein; Vacuolar protein sorting-associated protein 52-like protein-like protein; Vps52; VPS52 GARP complex subunit; VPS52 subunit of GARP complex; VPS52, GARP complex subunit; wu:fb94c11
Common Name VPS52
Gene Symbol VPS52
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DDSKEVESFQQLLNARTQEFIEELLSPPFGGLVAFVKEAEALIERGQAERLRGEEARVTQLIRGFGSSWKSSVESLSQDVMRSFTNFRNGTSIIQGA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.