missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VPS16 (aa 654-758) Control Fragment Recombinant Protein

Product Code. 30182473
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182473

Brand: Invitrogen™ RP99481

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83648 (PA5-83648. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps16 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2009]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H269
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64601
Name Human VPS16 (aa 654-758) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810074M16Rik; hVPS16; mVPS16; RP11-12M19.2; vacuolar protein sorting 16 (yeast); vacuolar protein sorting 16 homolog; vacuolar protein sorting 16 homolog (S. cerevisiae); vacuolar protein sorting protein 16; vacuolar protein sorting-associated protein 16 homolog; VPS16; VPS16 CORVET/HOPS core subunit; VPS16, CORVET/HOPS core subunit
Common Name VPS16
Gene Symbol VPS16
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AADAFYKAKNEFAAKATEDQMRLLRLQRRLEDELGGQFLDLSLHDTVTTLILGGHNKRAEQLARDFRIPDKRLWWLKLTALADLEDWEELEKFSKSKKSPIGYLP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.