missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Vitronectin, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17863733
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17863733 1 mg 1mg
17893203 100 μg 100µg
17873723 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17863733 Supplier Gibco™ Supplier No. AF140091MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Vitronectin is a 459 amino acid, single-chain, monomeric protein, which migrates at an apparent molecular weight of 75 kDa by SDS-PAGE under reducing conditions. The calculated molecular weight of Recombinant Human Vitronectin is 52.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04004
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7448
Molecular Weight (g/mol) 52.2 kDa
Name Human Vitronectin, Animal-Free
Quantity 1 mg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Common Name Vitronectin
Gene Symbol VTN
Biological Activity Recombinant human Vitronectin promotes attachment of hESC and iPSC in serum-free, feeder conditions at 5 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence DQESCKGRCT EGFNVDKKCQ CDELCSYYQS CCTDYTAECK PQVTRGDVFT MPEDEYTVYD DGEEKNNATV HEQVGGPSLT SDLQAQSKGN PEQTPVLKPE EEAPAPEVGA SKPEGIDSRP ETLHPGRPQP PAEEELCSGK PFDAFTDLKN GSLFAFRGQY CYELDEKAVR PGYPKLIRDV WGIEGPIDAA FTRINCQGKT YLFKGSQYWR FEDGVLDPDY PRNISDGFDG IPDNVDAALA LPAHSYSGRE RVYFFKGKQY WEYQFQHQPS QEECEGSSLS AVFEHFAMMQ RDSWEDIFEL LFWGRTSAGT RQPQFISRDW HGVPGQVDAA MAGRIYISGM APRPSLAKKQ RFRHRNRKGY RSQRGHSRGR NQNSRRPSRA TWLSLFSSEE SNLGANNYDD YRMDWLVPAT CEPIQSVFFF SGDKYYRVNL RTRRVDTVDP PYPRSIAQYW LGCPAPGHL
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.