missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VIPR2 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5416P-025
This item is not returnable.
View return policy
Description
VIPR2/VPAC2 Monoclonal specifically detects VIPR2/VPAC2 in Human samples. It is validated for Flow Cytometry.
Specifications
| VIPR2/VPAC2 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ16511, Helodermin-preferring VIP receptor, PACAP type III receptor, PACAPR3, PACAP-R3, PACAP-R-3, Pituitary adenylate cyclase-activating polypeptide type III receptor, vasoactive intestinal peptide receptor 2, vasoactive intestinal polypeptide receptor 2, VIP R2, VIP2R, VIP-R-2, VPAC2, VPCAP2R | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7434 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 476031 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P41587 | |
| VIPR2 | |
| NS0 mouse myeloma cell line transfected with human VIP R2 Glu24-Ile438 Accession # P41587 | |
| 25 Tests | |
| Primary | |
| Detects human VIP R2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction