missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VIP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6079
This item is not returnable.
View return policy
Description
VIP Monoclonal specifically detects VIP in Human samples. It is validated for Immunohistochemistry.
Specifications
| VIP | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7432 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 576721 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| VIP | |
| KLH-coupled human VIP synthetic peptides HSDAVFTDNYTR and KYLNSILN | |
| 100 μg | |
| Primary | |
| Detects recombinant human VIP in direct ELISAs. It does not detect the C-terminal region (aa 145-152). | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction