Learn More
Human Vimentin Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB21051-SP
 View more versions of this product
Product Code. 18792347
  • £168.00 / 25µg
Description
Vimentin Monoclonal specifically detects Vimentin in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vimentin
Monoclonal
LYOPH
Western Blot 1 ug/mL
P08670
VIM
E. coli-derived recombinant human Vimentin Ser2-Glu466 Accession # P08670
25 μg
Primary
Detects human Vimentin in direct ELISAs and Western blots.
Human
Purified
Western Blot
280622
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FLJ36605, VIM, vimentin
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
7431
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a