missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VIGR/GPR126 MAb (Clone 1044430), R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £393.00
Specifications
| Antigen | VIGR/GPR126 |
|---|---|
| Clone | 1044430 |
| Dilution | Flow Cytometry 0.25 ug/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762377
![]() |
R&D Systems
MAB10577-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763287
![]() |
R&D Systems
MAB10577-100 |
100 μg |
£393.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VIGR/GPR126 Monoclonal antibody specifically detects VIGR/GPR126 in Human samples. It is validated for Flow CytometrySpecifications
| VIGR/GPR126 | |
| Flow Cytometry 0.25 ug/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, Gm222, GPR126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, PS1TP2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2 | |
| Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437, Accession # AAH75798.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1044430 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAH75798.1 | |
| 57211 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title