missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VHL Antibody, R&D Systems™

Mouse Monoclonal Antibody
£206.00 - £469.00
Specifications
| Antigen | Von Hippel Lindau |
|---|---|
| Clone | 1066408 |
| Dilution | Immunocytochemistry 3-25 μg/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794474
![]() |
R&D Systems
MAB114352-100 |
100 μg |
£469.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18794324
|
R&D Systems
MAB114352-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Von Hippel Lindau Monoclonal antibody specifically detects Von Hippel Lindau in Human samples. It is validated for ImmunocytochemistrySpecifications
| Von Hippel Lindau | |
| Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
| E. coli -derived recombinant human VHL, Asn150-Asp213, Accession # P40337 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1066408 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P40337 | |
| 7428 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title