missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VHL Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB114351-SP
This item is not returnable.
View return policy
Description
Von Hippel Lindau Monoclonal antibody specifically detects Von Hippel Lindau in Human samples. It is validated for Immunohistochemistry
Specifications
| Von Hippel Lindau | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
| E. coli -derived recombinant human VHL, Asn150-Asp213, Accession # P40337 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 1066409 | |
| Immunohistochemistry 5-15 μg/mL | |
| P40337 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7428 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction