missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGFR3/Flt-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB349-500
This item is not returnable.
View return policy
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| VEGFR3/Flt-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2324 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 54716 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P35916 | |
| FLT4 | |
| Mouse myeloma cell line NS0-derived recombinant human VEGFR3, Tyr25-Ile776, Accession # P35916 | |
| 500 μg | |
| Primary | |
| Detects human VEGFR3 in ELISAs. In ELISAs, shows 25% cross-reactivity with recombinant mouse (rm) VEGFR3 and no cross-reactivity with rhVEGFR1 or rhVEGFR2. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction