missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGFR2/KDR/Flk-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£168.00 - £1540.00
Specifications
| Antigen | VEGFR2/KDR/Flk-1 |
|---|---|
| Clone | 89109 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743129
![]() |
R&D Systems
MAB3573-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784148
![]() |
R&D Systems
MAB3573-100 |
100 μg |
£389.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18735378
![]() |
R&D Systems
MAB3573-500 |
500 μg |
£1540.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| VEGFR2/KDR/Flk-1 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2 | |
| KDR | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 89109 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P35968 | |
| 3791 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VEGFR2/KDR/Flk-1 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) VEGFR1, rhVEGFR3, or recombinant mouse VEGFR2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title