missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGFR2/KDR/Flk-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
£190.00 - £352.00
Specifications
| Antigen | VEGFR2/KDR/Flk-1 |
|---|---|
| Clone | 89106 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758409
![]() |
R&D Systems
FAB357N-025 |
25 Tests |
£190.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18786667
![]() |
R&D Systems
FAB357N-100 |
100 Tests |
£352.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry.Specifications
| VEGFR2/KDR/Flk-1 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2 | |
| KDR | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human VEGF R2/KDR/Flk-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) VEGF R1, rhVEGF R3 or recombinant mouse VEGF R2 is observed. |
| 89106 | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Mouse | |
| Human | |
| P35968 | |
| 3791 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF R2/KDR/Flk-1 Ala20-Glu764 Accession # P35968 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title