missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human VEGFR1 Recombinant Protein
Description
This product contains purified recombinant human VeGF Receptor-1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
FLT1 (VEGFR1) is a receptor tyrosine kinase which plays a critical role in angiogenesis. Active angiogenesis has been found to play important role in many pathological and physiological processes, such as tumor progression, diabetic retinopathy, rheumatoid arthritis and wound healing. May play an essential role as a negative regulator of embryonic angiogenesis by inhibiting excessive proliferation of endothelial cells. Can promote endothelial cell proliferation, survival and angiogenesis in adulthood. Its function in promoting cell proliferation seems to be cell-type specific. Promotes PGF-mediated proliferation of endothelial cells, proliferation of some types of cancer cells, but does not promote proliferation of normal fibroblasts (in vitro). Has very high affinity for VEGFA and relatively low protein kinase activity; may function as a negative regulator of VEGFA signaling by limiting the amount of free VEGFA and preventing its binding to KDR.
Specifications
Specifications
| Accession Number | P17948 |
| Concentration | 0.29 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 25 mM sodium acetate with 50% glycerol and no preservative; pH 4.8 |
| Gene ID (Entrez) | 2321 |
| Name | Human VEGFR1 |
| pH Range | 4.8 |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?