missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF165b Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB3045-500
This item is not returnable.
View return policy
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
| VEGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7422 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 56-1 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunofluorescence | |
| AAL27435 | |
| VEGFA | |
| KLH-conjugated human VEGF 165b synthetic peptide TCRSLTRKD Accession # AAL27435 | |
| 500 μg | |
| Primary | |
| Detects human VEGF 165b in direct ELISAs and Western blots. Recognizes an epitope within a 9 amino acid sequence at the C-terminus of human VEGF 165b . In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) VEGF 206 rhVEGF-B 167 rhVEGF-B 186 rhVEGF-C, or rhVEGF-D is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction