missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF (Research Grade Bevacizumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
£197.00 - £456.00
Specifications
| Antigen | VEGF |
|---|---|
| Clone | Hu101 |
| Dilution | Neutralization 10-80 ng/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743607
![]() |
R&D Systems
MAB9947-SP |
25 μg |
£197.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772807
![]() |
R&D Systems
MAB9947-100 |
100 μg |
£456.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Neutralization.Specifications
| VEGF | |
| Neutralization 10-80 ng/mL | |
| Monoclonal | |
| Human | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 7422.0 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening. |
| Hu101 | |
| Neutralization | |
| Unconjugated | |
| Human | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Human VEGF-A | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VEGF based on Bevacizumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Bevacizumab. This product is for research use only. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title