missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VEGF Receptor 3/FLT4 ELISA Kit

Product Code. 15561577
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15561577

Brand: Invitrogen™ BMS2064TEN

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human VEGF-R3/FLT-4
Biotin-Conjugate anti-human VEGF-R3/FLT-4 monoclonal antibody
Streptavidin-HRP
Human VEGF-R3/FLT-4 Standard lyophilized, 80ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

ELISA

The Human Vascular Endothelial Growth Factor Receptor 3/FLT4 (Hu VEGFR3) ELISA quantitates Hu VEGFR3 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu VEGFR3. Principle of the method The Human VEGFR3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35916
Assay Range 0.6 to 40.0 ng/mL
Assay Sensitivity 0.03 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2324
Gene Symbol FLT-41,LMPH1A,PCL,VEGFR-3,VEGFR3,FLT4
Interassay CV 5%
Intraassay CV 5.7
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 20 μL, Serum 20 μL, Supernatant 20 μL
Storage Requirements 2°C to 8°C
Synonym FLT-4, VEGF receptor 3, PCL
Target VEGF Receptor 3
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.