Learn More
Invitrogen™ VEGFC Human ELISA Kit
For the quantitative detection of human VEGF-C.
Brand: Invitrogen™ BMS297-2TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human VEGF-C
Biotin-Conjugate anti-human VEGF-C monoclonal antibody
Streptavidin-HRP
Human VEGF-C Standard lyophilized, 15ng/mL upon reconstitution
Sample Diluent
Assay Diluent
Conjugate Diluent
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Vascular Endothelial Growth Factor-C (Hu VEGF-C) ELISA quantitates Hu VEGF-C in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu VEGF-C. Principle of the method The Human VEGF-C solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.Specifications
P49767 | |
0.057 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
Flt4-L,LMPH1D,VRP | |
6.7% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
2 hr. 30 min. |
0.23-15.0 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
7424 | |
12% | |
Pre-coated 96 well plate, Standard, Standard, Assay Buffer concentrate, Assay Buffer concentrate, SAV-HRP, SAV-HRP, Lysis Buffer, Lysis Buffer, Chromogen, Stop Solution | |
10 x 96 Tests | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
VRP, FLT-4 ligand | |
1 hr. 20 min. |
For Research Use Only