missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF-C Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 13 publications
Brand: R&D Systems AF752
This item is not returnable.
View return policy
Description
VEGF-C Polyclonal specifically detects VEGF-C in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| VEGF-C | |
| Polyclonal | |
| Unconjugated | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGFC, VEGF-C, VRPFLT4 ligand DHM | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7424 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA, Immunohistochemistry 5-15 ug/mL | |
| VEGFC | |
| E. coli-derived recombinant human VEGF-C | |
| 100 μg | |
| Primary | |
| Detects human VEGF-C in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction