missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF-C Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £1570.00
Specifications
| Antigen | VEGF-C |
|---|---|
| Clone | 193208 |
| Dilution | Western Blot 1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
18728388
![]() |
R&D Systems
MAB752-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18724239
![]() |
R&D Systems
MAB752-100 |
100 μg |
£389.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721717
![]() |
R&D Systems
MAB752-500 |
500 μg |
£1570.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
VEGF-C Monoclonal specifically detects VEGF-C in Human samples. It is validated for Western Blot, ELISA.Specifikationer
| VEGF-C | |
| Western Blot 1 ug/mL, ELISA | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q6FH59 | |
| 7424 | |
| Mouse myeloma cell line NS0-derived recombinant human VEGF-C Thr103-Arg227 (Cys156Ser) Accession # Q6FH59 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VEGF-C in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) PDGF-AA, rhPDGF-BB, rhPlGF, rhVEGF 165 rhVEGF 206 rhVEGF-B 186 or rhVEGF-D. |
| 193208 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGFC, VEGF-C, VRPFLT4 ligand DHM | |
| VEGFC | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel