missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF-B186 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB337
This item is not returnable.
View return policy
Description
VEGF-B Monoclonal specifically detects VEGF-B in Human samples. It is validated for Western Blot.
Specifications
| VEGF-B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| VEGFB | |
| E. coli-derived recombinant human VEGF-B 186 | |
| 500 μg | |
| Primary | |
| Detects human VEGF-B 186 in direct ELISAs and Western blots. In these formats, this antibody does not detect recombinant human (rh) VEGF-B 167 rmVEGF-B 186 rhVEGF 121 rhVEGF 186 rhVEGF 206 rhVEGF-C, rhVEGF-D, rhCTGF, rhPDGF, rhPlGF, or rhLDGF. | |
| Human | |
| Purified |
| Western Blot | |
| 67431 | |
| Western Blot 1 ug/mL | |
| vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7423 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction