missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VE-Cadherin Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£210.00 - £534.00
Specifications
| Antigen | VE-Cadherin |
|---|---|
| Clone | 1098251 |
| Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry, Western Blot, Simple Western |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30484801
|
R&D Systems
MAB11663-100 |
100 μg |
£534.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30484813
|
R&D Systems
MAB11663-SP |
25 μg |
£210.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Cadherin Monoclonal antibody specifically detects VE-Cadherin in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry,Western Blot,Simple WesternSpecifications
| VE-Cadherin | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
| Mouse myeloma cell line, NS0- derived recombinant human VE-Cadherin, Accession # P33151 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Antibody |
| 1098251 | |
| Immunohistochemistry, Immunocytochemistry, Western Blot, Simple Western | |
| Unconjugated | |
| Mouse | |
| Human | |
| P33151 | |
| 1003 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title