missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VDP (aa 553-634) Control Fragment Recombinant Protein

Product Code. 30213146
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213146

Brand: Invitrogen™ RP104687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60763
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8615
Name Human VDP (aa 553-634) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 115 kDa; General vesicular transport factor p115; P115; protein USO1 homolog; TAP; transcytosis associated protein; transcytosis associated protein p115; transcytosis-associated protein; USO1; USO1 homolog, vesicle docking protein; USO1 vesicle docking factor; USO1 vesicle docking protein homolog; USO1 vesicle transport factor; Vdp; VDP p115; vesicle docking protein p115; vesicle docking protein, 115 kDa; vesicle-docking protein
Common Name VDP
Gene Symbol USO1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DNSLESYMKEKLKQLIEKRIGKENFIEKLGFISKHELYSRASQKPQPNFPSPEYMIFDHEFTKLVKELEGVITKAIYKSSEE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.