missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human VDAC (aa 185-197) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Human VDAC1 (residues 185-197) Synthetic Peptide, PEP-082, from Invitrogen.
Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
Specifications
Specifications
| Gene ID (Entrez) | 7416 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P21796 |
| Common Name | Human VDAC (aa 185-197) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction