missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VCY1B (aa 55-83) Control Fragment Recombinant Protein

Product Code. 30207102
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207102

Brand: Invitrogen™ RP104377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60888 (PA5-60888. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of a family of human VCX/Y genes. This gene family has multiple members on both X and Y chromosomes, and all are expressed exclusively in male germ cells. Members of the VCX/Y family share a high degree of sequence identity, with the exception that a 30-bp unit is tandemly repeated in X-linked members but occurs only once in Y-linked members. VCX/Y genes encode small and highly charged proteins of unknown function. This gene encodes a small, positively charged protein. The presence of a putative bipartite nuclear localization signal suggests that this gene encodes a nuclear protein. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number O14598
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9084
Name Human VCY1B (aa 55-83) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Basic charge, Y-linked 1; basic protein on Y chromosome, 1; BPY1; BPY1B; testis-specific basic protein on Y, 1; testis-specific basic protein Y 1; variable charge, Y chromosome; variable charge, Y-linked; variably charged protein Y; VCY; VCY1; VCY1A; VCY1B
Common Name VCY1B
Gene Symbol VCY
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AATKMAAVTAPEAESGPAAPGPSDQPSQE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.