missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VCAM-1/CD106 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £430.00
Specifications
| Antigen | VCAM-1/CD106 |
|---|---|
| Clone | 1027433 |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720097
![]() |
R&D Systems
MAB8091-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791987
![]() |
R&D Systems
MAB8091-100 |
100 μg |
£430.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| VCAM-1/CD106 | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Chinese Hamster Ovary cell line CHO-derived human VCAM-1/CD106, Phe25-Glu698, Accession # P19320-1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1027433 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P19320-1 | |
| 7412 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title