missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VCAM-1/CD106 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8092-100
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Chinese Hamster Ovary cell line CHO-derived human VCAM-1/CD106, Phe25-Glu698, Accession # P19320-1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 2691A | |
| Western Blot 4 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
| P19320-1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7412 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction