missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VCAM-1/CD106 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF809-SP
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Polyclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Simple Western, Adhesion Blockade.
Specifications
| VCAM-1/CD106 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| VCAM1 | |
| Chinese hamster ovary cell line (CHO)-derived recombinant human VCAM-1 Extracellular domain | |
| 25 μg | |
| Primary | |
| Detects VCAM-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse VCAM-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Blocking Assay | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Adhesion Blockade | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7412 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction