missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VASA Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 22 publications
Brand: R&D Systems AF2030-SP
This item is not returnable.
View return policy
Description
VASA Polyclonal specifically detects VASA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VASA | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DDX4, DEAD (Asp-Glu-Ala-Asp) box polypeptide 4, DEAD box protein 4, EC 3.6.1, EC 3.6.4.13, MGC111074, probable ATP-dependent RNA helicase DDX4, Vasa homolog, VASADEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54514 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9NQI0 | |
| DDX4 | |
| E. coli-derived recombinant human VASA Met1-Tyr145 Accession # Q9NQI0 | |
| 25 μg | |
| Primary | |
| Detects human VASA in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction