missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Human VARS2 (aa 751-823) Control Fragment Recombinant Protein
Klicka för att se tillgängliga alternativ
Quantity:
100 μL
Packungsgröße:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-66493 (PA5-66493, PA5-64437 (PA5-64437. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a mitochondrial aminoacyl-tRNA synthetase, which catalyzes the attachment of valine to tRNA(Val) for mitochondrial translation. Mutations in this gene cause combined oxidative phosphorylation deficiency-20, and are also associated with early-onset mitochondrial encephalopathies. Alternative splicing of this gene results in multiple transcript variants.
Spezifikation
Spezifikation
| Accession Number | Q5ST30 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57176 |
| Name | Human VARS2 (aa 751-823) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1190004I24Rik; COXPD20; Kiaa1885; mKIAA1885; valine tRNA ligase 2, mitochondrial (putative); valine--tRNA ligase, mitochondrial; ValRS; valyl trna synthetase; Valyl-tRNA synthetase; valyl-tRNA synthetase 2, mitochondrial; valyl-tRNA synthetase 2, mitochondrial (putative); valyl-tRNA synthetase like; valyl-tRNA synthetase, mitochondrial; Valyl-tRNA synthetase-like; VARS2; VARS2L; VARSL |
| Common Name | VARS2 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur