missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human VAP-1 Recombinant Protein, PeproTech®

Product Code. 17841243
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17841243

Brand: Gibco™ 15016250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human VAP-1 is a mixture of monomeric and disulfide-linked homodimeric forms of a 737 amino acid polypeptide, corresponding to amino acids 27 to 763 of the VAP-1 precursor. The calculated molecular weight of Recombinant Human VAP-1 is 81.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16853
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 8639
Molecular Weight (g/mol) 81.8 kDa
Name Human VAP-1
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97
Common Name AOC3
Gene Symbol AOC3
Biological Activity Measured by its ability to produce hydrogen peroxide during the oxidation of benzylamine. The specific activity >16 pMoles/min/ug of VAP-1.
Conjugate Unconjugated
Recombinant Recombinant
Sequence GRGGDGGEPS QLPHCPSVSP SAQPWTHPGQ SQLFADLSRE ELTAVMRFLT QRLGPGLVDA AQARPSDNCV FSVELQLPPK AAALAHLDRG SPPPAREALA IVFFGRQPQP NVSELVVGPL PHPSYMRDVT VERHGGPLPY HRRPVLFQEY LDIDQMIFNR ELPQASGLLH HCCFYKHRGR NLVTMTTAPR GLQSGDRATW FGLYYNISGA GFFLHHVGLE LLVNHKALDP ARWTIQKVFY QGRYYDSLAQ LEAQFEAGLV NVVLIPDNGT GGSWSLKSPV PPGPAPPLQF YPQGPRFSVQ GSRVASSLWT FSFGLGAFSG PRIFDVRFQG ERLVYEISLQ EALAIYGGNS PAAMTTRYVD GGFGMGKYTT PLTRGVDCPY LATYVDWHFL LESQAPKTIR DAFCVFEQNQ GLPLRRHHSD LYSHYFGGLA ETVLVVRSMS TLLNYDYVWD TVFHPSGAIE IRFYATGYIS SAFLFGATGK YGNQVSEHTL GTVHTHSAHF KVDLDVAGLE NWVWAEDMVF VPMAVPWSPE HQLQRLQVTR KLLEMEEQAA FLVGSATPRY LYLASNHSNK WGHPRGYRIQ MLSFAGEPLP QNSSMARGFS WERYQLAVTQ RKEEEPSSSS VFNQNDPWAP TVDFSDFINN ETIAGKDLVA WVTAGFLHIP HAEDIPNTVT VGNGVGFFLR PYNFFDEDPS FYSADSIYFR GDQDAGACEV NPLACLPQAA ACAPDLPAFS HGGFSHN
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.