missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VAP-1/AOC3 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC39571N-100UG
This item is not returnable.
View return policy
Description
VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| VAP-1/AOC3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q16853 | |
| AOC3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VAP-1/AOC3 Gly27-Asn763 Accession # Q16853 | |
| RUO | |
| 8639 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 393106 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| amine oxidase, copper containing 3 (vascular adhesion protein 1), AOC3, Copper amine oxidase, EC 1.4.3, HPAO, HPAOSSAO, Semicarbazide-sensitive amine oxidase, SSAO, VAP1, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human VAP-1/AOC3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse VAP-1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction