missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VAMP-5 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7366-SP
This item is not returnable.
View return policy
Description
VAMP-5 Polyclonal specifically detects VAMP-5 in Human samples. It is validated for Immunohistochemistry.
Specifications
| VAMP-5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| myobrevin, VAMP5, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10791 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| O95183 | |
| VAMP5 | |
| E. coli-derived recombinant human VAMP-5 Ala2-Cys72 Accession # O95183 | |
| 25 μg | |
| Primary | |
| Detects human VAMP-5 in direct ELISAs. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) VAMP-7 is observed, and less than 1% cross-reactivity with rhVAMP-1, rhVAMP-2, and rhVAMP-8 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction