missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human V-ATPase H (aa 404-474) Control Fragment Recombinant Protein

Product Code. 30205366
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205366

Brand: Invitrogen™ RP93733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54793 (PA5-54793. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular organelles. V-ATPase-dependent organelle acidification is necessary for multiple processes including protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. The encoded protein is the regulatory H subunit of the V1 domain of V-ATPase, which is required for catalysis of ATP but not the assembly of V-ATPase. Decreased expression of this gene may play a role in the development of type 2 diabetes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UI12
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51606
Name Human V-ATPase H (aa 404-474) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0710001F19Rik; Atp6v1h; ATPase H+ transporting V1 subunit H; ATPase, H+ transporting, lysosomal 50/57 kDa, V1 subunit H; ATPase, H+ transporting, lysosomal V1 subunit H; AU022349; CGI-11; MSTP042; NBP1; Nef-binding protein 1; protein VMA13 homolog; SFD; SFDalpha; SFDbeta; vacuolar ATP synthase subunit H; vacuolar ATPase subunit H; vacuolar proton pump H subunit; vacuolar proton pump subunit H; Vacuolar proton pump subunit SFD; V-ATPase 50/57 kDa subunits; V-ATPase H subunit; V-ATPase subunit H; VMA13; V-type proton ATPase subunit H
Common Name V-ATPase H
Gene Symbol ATP6V1H
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PQVLAVAAHDVGEYVRHYPRGKRVIEQLGGKQLVMNHMHHEDQQVRYNALLAVQKLMVHNWEYLGKQLQSE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.