missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human V-ATPase G1 (aa 42-115) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65342 (PA5-65342. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation.
Specifications
Specifications
| Accession Number | O75348 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9550 |
| Name | Human V-ATPase G1 (aa 42-115) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1810024D14Rik; AA960677; ATP6G; ATP6G1; ATP6GL; ATP6J; ATP6V1G1; ATPase H+ transporting V1 subunit G1; ATPase, H transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J; ATPase, H+ transporting, lysosomal 13 kD, V1 subunit G; ATPase, H+ transporting, lysosomal 13 kDa, V1 subunit G1; ATPase, H+ transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, V1 subunit G; DKFZp547P234; lysosomal 13 kDa; vacuolar ATP synthase subunit M16; vacuolar H(+)-ATPase subunit G 1; vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16; VAG1; V-ATPase 13 kDa subunit 1; V-ATPase subunit G 1; Vma10; V-type proton ATPase subunit G 1 |
| Common Name | V-ATPase G1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction