missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human USP8 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7735
This item is not returnable.
View return policy
Description
UBPY/USP8 Polyclonal specifically detects UBPY/USP8 in Human samples. It is validated for Western Blot.
Specifications
| UBPY/USP8 | |
| Polyclonal | |
| Unconjugated | |
| P40818 | |
| USP8 | |
| E. coli-derived recombinant human USP8 Met1-Thr1118 Accession # P40818 | |
| 100 μg | |
| Primary | |
| Detects human USP8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human USP2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Deubiquitinating enzyme 8, EC 3.1.2.15, EC 3.4.19.12, HumORF8, KIAA0055hUBPy, MGC129718, ubiquitin carboxyl-terminal hydrolase 8, Ubiquitin isopeptidase Y, ubiquitin specific peptidase 8, ubiquitin specific protease 8, ubiquitin thioesterase 8, Ubiquitin thiolesterase 8, Ubiquitin-specific-processing protease 8, UBPY, UBPYFLJ34456 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9101 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction