missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human USP42 (aa 454-580) Control Fragment Recombinant Protein

Product Code. 30199478
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199478

Brand: Invitrogen™ RP89246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52338 (PA5-52338. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

USP42 (ubiquitin specific peptidase 42) is a 1,325 amino acid protein that belongs to the peptidase C19 family. Expressed in a variety of tissues, USP42 functions to catalyze the conversion of a ubiquitin C-terminal thioester to a free ubiquitin and a thiol. The gene encoding USP42 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H9J4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84132
Name Human USP42 (aa 454-580) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410140K03Rik; 3110031A07Rik; A630018G05Rik; D5Ertd591e; Deubiquitinating enzyme 42; ubiquitin carboxyl-terminal hydrolase 42; ubiquitin specific peptidase 42; ubiquitin specific protease 42; ubiquitin thioesterase 42; ubiquitin thiolesterase 42; ubiquitin-specific-processing protease 42; Usp42
Common Name USP42
Gene Symbol Usp42
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PQLPSHMIKNPPHLNGTGPLKDTPSSSMSSPNGNSSVNRASPVNASASVQNWSVNRSSVIPEHPKKQKITISIHNKLPVRQCQSQPNLHSNSLENPTKPVPSSTITNSAVQSTSNASTMSVSSKVTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.