missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human USP37 (aa 17-135) Control Fragment Recombinant Protein

Product Code. 30212555
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212555

Brand: Invitrogen™ RP89265

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Deubiquitinase that antagonizes the anaphase-promoting complex (APC/C) during G1/S transition by mediating deubiquitination of cyclin-A (CCNA1 and CCNA2), thereby promoting S phase entry. Specifically mediates deubiquitination of 'Lys-11'-linked polyubiquitin chains, a specific ubiquitin-linkage type mediated by the APC/C complex. Also mediates deubiquitination of 'Lys-48'-linked polyubiquitin chains in vitro. Phosphorylation at Ser-628 during G1/S phase maximizes the deubiquitinase activity, leading to prevent degradation of cyclin-A (CCNA1 and CCNA2) (PubMed:21596315). Plays an important role in the regulation of DNA replication by stabilizing the licensing factor CDT1 (PubMed:27296872). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86T82
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57695
Name Human USP37 (aa 17-135) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4932415L06Rik; C330008N13Rik; Deubiquitinating enzyme 37; Kiaa1594; tmp_locus_50; ubiquitin carboxyl-terminal hydrolase 37; ubiquitin specific peptidase 37; ubiquitin specific peptidase 37-like; ubiquitin specific protease 37; ubiquitin thioesterase 37; ubiquitin thiolesterase 37; ubiquitin-specific-processing protease 37; USP37
Common Name USP37
Gene Symbol Usp37
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TGITKWKEGSFEIVEKENKVSLVVHYNTGGIPRIFQLSHNIKNVVLRPSGAKQSRLMLTLQDNSFLSIDKVPSKDAEEMRLFLDAVHQNRLPAAMKPSQGSGSFGAILGSRTSQKETSR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.