missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit

Product Code. 30000869
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Unit Size:
1 set
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000869 1 Pack (6 Plates) Pack of 96
18761964 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30000869 Supplier R&D Systems™ Supplier No. SKM100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TIM-1/KIM-1/HAVCR in urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.046 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Urine
Specificity Natural and recombinant human TIM-1
For Use With (Application) ELISA
Gene ID (Entrez) 26762
Gene Symbol HAVCR1
Quantity 1 Pack (6 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Target TIM-1/KIM-1/HAVCR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.