missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human UQCRQ (aa 3-41) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61285 (PA5-61285. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain.
Specifications
Specifications
| Accession Number | O14949 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 27089 |
| Name | Human UQCRQ (aa 3-41) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1100001F06Rik; 1500040F11Rik; 5830407L17Rik; 9.5 kDa; AA959903; c1502; Complex III subunit 8; complex III subunit VII; complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5 kD); low molecular mass ubiquinone-binding protein (Qpc-pending); MC3DN4; QCR8; QPC; QP-C; transcript under zygotic control; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5 kDa; unknown; UQCR7; Uqcrb; UQCRQ |
| Common Name | UQCRQ |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction