missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human uPAR Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
£184.00 - £1790.00
Specifications
| Antigen | uPAR |
|---|---|
| Clone | 62022 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732959
![]() |
R&D Systems
MAB807-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787758
![]() |
R&D Systems
MAB807-100 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706829
![]() |
R&D Systems
MAB807-500 |
500 μg |
£1790.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
uPAR Monoclonal specifically detects uPAR in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| uPAR | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q03405 | |
| 5329 | |
| Mouse myeloma cell line NS0-derived recombinant human uPAR Leu23-Arg303 Accession # Q03405 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human uPAR in direct ELISAs and Western blots. When used in a sandwich ELISA in combination with the biotinylated anti-human uPAR detection antibody (Catalog # BAF807), no significant cross-reactivity was observed with recombinant mouse uPAR. |
| 62022 | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, PLAUR, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| PLAUR | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title