missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Description
The Human Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Universal |
| Quantity | 1 Set |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Universal Protein Lysate: Human Normal Tissues |
| Cellular Fraction | Whole |
| Tissue Condition | Normal |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Afficher plus |
For Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu