missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-2/5/6 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1298S-100UG
This item is not returnable.
View return policy
Description
ULBP-2/5/6 Monoclonal specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry.
Specifications
| ULBP-2/5/6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BZM5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. It does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 165903 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ALCAN-alpha, N2DL2, NKG2DL2, RAET1G, RAET1H, RAET1L, UL16 binding protein 2, ULBP2, ULBP5, ULBP6 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-2 Accession # Q9BZM5 | |
| RUO | |
| 80328 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction