missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UGT8 (aa 205-311) Control Fragment Recombinant Protein

Product Code. 30195091
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195091

Brand: Invitrogen™ RP107455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111606 (PA5-111606. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16880
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7368
Name Human UGT8 (aa 205-311) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2-hydroxyacylsphingosine 1-beta-galactosyltransferase; AI850488; AW455908; ceramide UDP-galactosyltransferase; Cerebroside synthase; Cgt; mCerGT; UDP galactosyltransferase 8; UDP galactosyltransferase 8 A; UDP glycosyltransferase 8; UDP-galactose ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase 8; UDP-galactose-ceramide galactosyltransferase 8 A; UDP-glucuronosyltransferase 8; Ugt4; UGT8; Ugt8a; uridine diphosphate glycosyltransferase 8
Common Name UGT8
Gene Symbol Ugt8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LGVSFLVLPKYERIMQKYNLLPEKSMYDLVHGSSLWMLCTDVALEFPRPTLPNVVYVGGILTKPASPLPEDLQRWVNGANEHGFVLVSFGAGVKYLSEDIANKLAGA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.