missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UGT2B4 (aa 276-300) Control Fragment Recombinant Protein

Product Code. 30197938
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197938

Brand: Invitrogen™ RP104381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60926 (PA5-60926. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

UGT2B4 belongs to the UDP-glycosyltransferase family. UDPGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. This isozyme is active on polyhydroxylated estrogens (such as estriol, 4-hydroxyestrone and 2-hydroxyestriol) and xenobiotics (such as 4-methylumbelliferone, 1-naphthol, 4-nitrophenol, 2-aminophenol, 4-hydroxybiphenyl and menthol). It is capable of 6 alpha-hydroxyglucuronidation of hyodeoxycholic acid.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06133
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7363
Name Human UGT2B4 (aa 276-300) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HLUG25; hyodeoxycholic acid; hyodeoxycholic acid-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B15; UDP glucuronosyltransferase 2 family, polypeptide B36; UDP glucuronosyltransferase 2 family, polypeptide B4; UDP glucuronosyltransferase family 2 member B4; UDP glycosyltransferase 2 family, polypeptide B4; UDP-glucuronosyltransferase 2B12; UDP-glucuronosyltransferase 2B15; UDP-glucuronosyltransferase 2B36; UDP-glucuronosyltransferase 2B4; UDP-glucuronyltransferase, family 2, beta-4; UDPGT 2B12; UDPGT 2B15; UDPGT 2B36; UDPGT 2B4; UDPGT2B4; UDPGTH1; UDPGTh-1; UGT2B11; Ugt2b12; Ugt2b15; Ugt2b36; UGT2B4
Common Name UGT2B4
Gene Symbol UGT2B4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FVGGLHCKPAKPLPKEMEEFVQSSG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.