missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human UGRP1/SCGB3A2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3545
This item is not returnable.
View return policy
Description
UGRP1/SCGB3A2 Polyclonal specifically detects UGRP1/SCGB3A2 in Human samples. It is validated for Western Blot.
Specifications
| UGRP1/SCGB3A2 | |
| Polyclonal | |
| Unconjugated | |
| Q96PL1 | |
| SCGB3A2 | |
| E. coli-derived recombinant human UGRP1/SCGB3A2 Phe22-Val93 Accession # Q96PL1 | |
| 100 μg | |
| Primary | |
| Detects human UGRP1/SCGB3A2 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse UGRP1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| LU103, Pneumo secretory protein 1, PnSP-1, PNSP1secretoglobin family 3A member 2, SCGB3A2, secretoglobin, family 3A, member 2, UGRP1pnSP-1, Uteroglobin-related protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 117156 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction