missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UCMA (aa 89-138) Control Fragment Recombinant Protein

Product Code. 30199723
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199723

Brand: Invitrogen™ RP100763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61295 (PA5-61295. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

UCMA is a gene that encodes for a protein that is specific to chondrocytes and highly charged. This protein is expressed abundantly in the upper immature zone of fetal and juvenile epiphyseal cartilage. The encoded protein undergoes proteolytic processing to generate a mature protein that is secreted into the extracellular matrix. The glutamic acid residues in the protein undergo gamma carboxylation in a vitamin K-dependent manner. Undercarboxylation of the protein has been associated with osteoarthritis in humans. Alternative splicing of the gene results in multiple transcript variants that encode different isoforms. UCMA is also associated with diseases such as Osteoarthritis and Poland Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WVF2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 221044
Name Human UCMA (aa 89-138) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110017I16Rik; AW121955; C10orf49; Gla-rich protein; GRP; GRP/UCMA; RGD1308977; Ucma; ucma {ECO:0000250; Ucma-C; UniProtKB:Q14BU0}; Unique cartilage matrix-associated protein; unique cartilage matrix-associated protein {ECO:0000250; Unique cartilage matrix-associated protein C-terminal fragment; upper zone of growth plate and cartilage matrix associated; upper zone of growth plate and cartilage matrix associated protein
Common Name UCMA
Gene Symbol UCMA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YYEEQRNEFENFVEEQNDEQEERSREAVEQWRQWHYDGLHPSYLYNRHHT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.